Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FluorEver

FluorEver is a mounting medium which significantly extends a light-emitting capacity of fluorescent dyes which tend to fade during storage and microscopy examination.

$113.85 - $171.35

Specifications

Product Type Slide mounting media
Applications Fluorescence imunohistochemistry and cytochemistry, fluorescence neuronal tracing, and gene expression
Storage Requirements Store at −20°C in the dark. Avoid freeze-thaw cycles.
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
NB602295ML
View Documents
Novus Biologicals™
NBP2602295ML
5mL
Each for $113.85
Add to cart
 
NB6022910ML
View Documents
Novus Biologicals™
NBP26022910ML
10mL
Each for $171.35
Add to cart
 
Description

Description

  • Can be used for various applications including, but not limited to, fluorescence imunohistochemistry and cytochemistry, fluorescence neuronal tracing, and gene expression studies (using, for example, GFP and YFP tags)
  • Compatible with cryostat and paraffin-embedded tissue sections, as well as with cultured neuronal and non-neuronal cells.
  • Protects cells and tissues from shrinking thus maintaining their native cytological and histological architecture, which is of critical importance for accurate image analysis studies
  • Blocks fading of a large variety of fluorescent dyes within a broad range of excitation and emission spectra, which makes this mounting medium a reagent of choice for maintaining brightness of multi-color fluorescence samples during storage and analysis by either conventional fluorescence or laser confocal microscopy
Specifications

Specifications

Slide mounting media
Fluorescence imunohistochemistry and cytochemistry, fluorescence neuronal tracing, and gene expression
Store at −20°C in the dark. Avoid freeze-thaw cycles.
SDS

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit