Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride Standard, 1 ppm F Premixed with TISAB (50/50), Color Coded Green, Reagents

Catalog No. 01335202
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Catalog No. Quantity
01-335-202 4 L
01-335-205 10 L
01-335-204 1 L
01-335-203 20 L
01-335-201 500 mL
5 options

Catalog No. 01-335-202

Supplier: Reagents Holdings Llc CF1353TG4A

Only null left
Add to Cart
Add to Cart

Fluoride Standard, 1 ppm F Premixed with TISAB (50/50), Color Coded Green, Reagent, use for Water analysis (Fluoride content). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Fluoride Standard, 1 ppm F Premixed with TISAB (50/50), Color Coded Green
Packaging Poly Bottle
CAS 7732-18-5, 64-19-7, 7647-14-5
Specific Gravity 1.05
Physical Form Liquid
Color Green
pH 5 to 5.5
Boiling Point 100°C
For Use With (Equipment) Fluoride Meter
Solubility Completely miscible
Quantity 4 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.