Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride Standard, 10 ppm F Premixed with TISAB / Color Coded Orange, Reagents

Catalog No. 01335226
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L

Fluoride Standard, 10 ppm F Premixed with TISAB / Color Coded Orange, Reagent, use for Water analysis (Fluoride content). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Fluoride Standard, 10 ppm F Premixed with TISAB / Color Coded Orange
Packaging Poly Bottle
CAS 7732-18-5, 64-19-7, 7647-14-5
Specific Gravity 1.05
Physical Form Liquid
Color Orange
pH 5 to 5.5
Boiling Point 100°C
For Use With (Equipment) Fluoride Meter
Solubility Completely miscible
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.