Learn More
BioVision IncSupplier Diversity Partner CaspGLOW™ Fluorescein Active Caspase-3 Staining Kit: Convenient & Sensitive Kit to Detect Activated Caspases-3 in Living Cells. Detection Method: Fluorescence Microscopy, Flow Cytometry or Fluorescence Plate Reader. Kit Size: 25 and 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K18325
Activation of caspases plays a central role in apoptosis. The CaspGLOW™ Fluorescein Active Caspase-3 Staining Kit provides a convenient means for sensitive detection of activated caspase-3 in living cells. The assay utilizes the caspase-3 inhibitor, DEVD-FMK, conjugated to FITC (FITC-DEVD-FMK) as a marker. FITC-DEVD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-3 in apoptotic cells. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9593