Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
164298-25-3
C9H16FN2·F6P
316.2
MNJUGQKOHJQOCK-UHFFFAOYSA-N
btffh,bis tetramethylene fluoroformamidinium hexafluorophosphate,fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate,c9h16fn2.f6p,fluoro-dipyrrolidinocarbenium hexafluorophosphate,1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v,1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate,fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t,1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate,fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate
10935980
1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F
164298-25-3
C9H16FN2·F6P
316.2
MNJUGQKOHJQOCK-UHFFFAOYSA-N
btffh,bis tetramethylene fluoroformamidinium hexafluorophosphate,fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate,c9h16fn2.f6p,fluoro-dipyrrolidinocarbenium hexafluorophosphate,1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v,1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate,fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t,1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate,fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate
10935980
1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F
Coupling reagent
Melting Point 152.0°C to 154.0°C
Color White
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Elemental Analysis 32.82 to 35.55% C, 8.51 to 9.21% N
Assay Percent Range 96% min. (HPLC)
Formula Weight 316.2
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material Fluoro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning