Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoroacetone, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
430-51-3
C3H5FO
76.06
MFCD00000451
MSWVMWGCNZQPIA-UHFFFAOYSA-N
fluoroacetone,1-fluoro-2-propanone,1-fluoroacetone,2-propanone, 1-fluoro,mono-fluoroacetone,acetone, fluoro,1-fluoroacetone #,ch3coch2f,acmc-1bne0
9889
1-fluoropropan-2-one
CC(=O)CF
430-51-3
C3H5FO
76.06
MFCD00000451
MSWVMWGCNZQPIA-UHFFFAOYSA-N
fluoroacetone,1-fluoro-2-propanone,1-fluoroacetone,2-propanone, 1-fluoro,mono-fluoroacetone,acetone, fluoro,1-fluoroacetone #,ch3coch2f,acmc-1bne0
9889
1-fluoropropan-2-one
CC(=O)CF
Specific Gravity 1.054
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 7°C
Color Red
Boiling Point 75.0°C
Refractive Index 1.3660 to 1.3700
Quantity 1g
Density 1.0540g/mL
Assay Percent Range 97%
Linear Formula CH3COCH2F
Formula Weight 76.06
Physical Form Powder
Percent Purity 97%
Chemical Name or Material Fluoroacetone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger