Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoroboric acid, 50 wt.% solution in water, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Plastic bottle
16872-11-0,7732-18-5,10043-35-3
BF4H
87.812
MFCD00011345
ODGCEQLVLXJUCC-UHFFFAOYSA-O
fluoroboric acid,fluoboric acid,tetrafluoroboric acid,hydrogen tetrafluoroborate,borofluoric acid,hydrofluoboric acid,borate 1-, tetrafluoro-, hydrogen,hydron tetrafluoroborate,hydrogen tetrafluoroborate 1-,unii-h429wz9fbq
28118
CHEBI:38902
hydron;tetrafluoroborate
[H+].[B-](F)(F)(F)F
16872-11-0,7732-18-5,10043-35-3
BF4H
87.812
MFCD00011345
ODGCEQLVLXJUCC-UHFFFAOYSA-O
fluoroboric acid,fluoboric acid,tetrafluoroboric acid,hydrogen tetrafluoroborate,borofluoric acid,hydrofluoboric acid,borate 1-, tetrafluoro-, hydrogen,hydron tetrafluoroborate,hydrogen tetrafluoroborate 1-,unii-h429wz9fbq
28118
CHEBI:38902
hydron;tetrafluoroborate
[H+].[B-](F)(F)(F)F
C-X bond formation reagent
Quantity 25g
Color White
Linear Formula HBF4
UN Number 1775
Fieser 01, 394
Merck Index 15, 4180
Chloride 50ppm max.
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol
Density 1.4100g/mL
Formula Weight 87.81
Physical Form Adhering Crystals and/or Chunks
Percent Purity 48 to 51% (Acid)
Fluorosilicic Acid 200ppm (H2SiF6) max.
Free Acid 2.5% max. (free boric acid)
Iron (Fe) 50ppm max.
Lead (Pb) 500ppm max.
Packaging Plastic bottle
Solubility Solubility in water: miscible. Other solubilities: miscible with alcohol
Specific Gravity 1.41
Storage Information May develop some turbidity or precipitate
Sulfate 200ppm max.
Melting Point -90.0°C
Boiling Point 130.0°C
Chemical Name or Material Fluoroboric acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 240-898-3

RTECSNumber : ED2685000

TSCA : TSCA