Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorodimethylphenylsilane 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass Bottle
454-57-9
C8H11FSi
154.259
MFCD03093772
OCKMVMKNCKNCMZ-UHFFFAOYSA-N
dimethylphenylfluorosilane,fluorodimethylphenylsilane,silane, fluorodimethylphenyl,fluorodimethyl phenyl silane,silane,fluorodimethylphenyl,benzene, fluorodimethylsilyl,ockmvmknckncmz-uhfffaoysa
9962
fluoro-dimethyl-phenylsilane
C[Si](C)(C1=CC=CC=C1)F
454-57-9
C8H11FSi
154.259
MFCD03093772
OCKMVMKNCKNCMZ-UHFFFAOYSA-N
dimethylphenylfluorosilane,fluorodimethylphenylsilane,silane, fluorodimethylphenyl,fluorodimethyl phenyl silane,silane,fluorodimethylphenyl,benzene, fluorodimethylsilyl,ockmvmknckncmz-uhfffaoysa
9962
fluoro-dimethyl-phenylsilane
C[Si](C)(C1=CC=CC=C1)F
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4730 to 1.4750
Quantity 5g
Color Colorless
Boiling Point 42.0°C to 46.0°C (20.0mmHg)
Assay Percent Range 97%
Linear Formula (CH3)2Si(F)C6H5
Formula Weight 154.26
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Fluorodimethylphenylsilane
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger