Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Fluorolink™ E10-H

Click to view available options
Quantity:
25g
100g
162492-15-1
162492-15-1

Use in polymer modifications

Applications
Use in polymer modifications.

Solubility
Soluble in fluorinated solvents, alcohol, and ketones. Insoluble in water.

Notes
Incompatible with heat and oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Quantity 100g
Density 1.73g/mL
Odor Odorless
Color Colorless to Yellow
Refractive Index 1.3170
Solubility Information Soluble in fluorinated solvents, alcohol, and ketones; Insoluble in water
Physical Form Liquid
Chemical Name or Material Fluorolink™ E10-H