Alfa Aesar™ Fluorophosphoric acid, 70% w/w aq. soln.

Manufacturer: Alfa Aesar™ H2622122

Quantity 100g

 View more versions of this product

Catalog No. AAH2622122

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13537-32-1
    Molecular Formula FP(O)(OH)2
    Formula Weight 99.99
    MDL Number MFCD00011334
    UN Number UN1776
    Chemical Name or Material Fluorophosphoric acid

    Specifications

    Formula Weight 99.99
    Concentration 70% w/w aq. soln.
    MDL Number MFCD00011334
    UN Number UN1776
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FLUOROPHOSPHORIC ACID, ANHYDROUS
    Quantity 100g
    Molecular Formula FP(O)(OH)2
    CAS 13537-32-1
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Fluorophosphoric acid

    GHS H Statement
    H300-H310-H330-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit