Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorouracil, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
51-21-8
C4H3FN2O2
130.08
GHASVSINZRGABV-UHFFFAOYSA-N
5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
FC1=CNC(=O)NC1=O
51-21-8
C4H3FN2O2
130.08
GHASVSINZRGABV-UHFFFAOYSA-N
5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
FC1=CNC(=O)NC1=O

FL127, 51-21-8, C4H3FN2O2

Spectrum™ Fluorouracil, USP belongs to a class of medications known as anti-metabolites that treat pre-cancerous and cancerous skin growths. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 100 g
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 102%