Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoxetine hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

56296-78-7
C17H19ClF3NO
345.79
GIYXAJPCNFJEHY-UHFFFAOYSA-N
N-Methyl-3-[(4-trifluoromethyl)phenoxy]-3-phenylpropylamine hydrochloride
62857

5-HT reuptake inhibitor

Selective serotonin reuptake inhibitor. Binds to the human 5-HT transporter with a Ki of 0.9 nmol/l and is between 150- and 900- fold selective over 5-HT1A, 5-HT2A, H1, α1, α2-adrenergic, and muscarinic receptors. Antidepressant. Induces differentiation of neuronal precursors, enhancing neuronal characteristics.
Quantity 10mg
SMILES CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Formula Weight 345.78
Chemical Name or Material Fluoxetine hydrochloride
Grade >99%

Recommended Storage : Store at Room Temperature