Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoxetine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
56296-78-7
C17H19ClF3NO
345.79
GIYXAJPCNFJEHY-UHFFFAOYNA-N
hydrogen methyl({3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl})amine chloride
[H+].[Cl-].CNCCC(OC1=CC=C(C=C1)C(F)(F)F)C1=CC=CC=C1
56296-78-7
C17H19ClF3NO
345.79
GIYXAJPCNFJEHY-UHFFFAOYNA-N
hydrogen methyl({3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl})amine chloride
[H+].[Cl-].CNCCC(OC1=CC=C(C=C1)C(F)(F)F)C1=CC=CC=C1

F1200, 56296-78-7, C17H18F3NO•HCl

Spectrum™ Fluoxetine Hydrochloride, USP is used for the treatment of major depressive disorder, obsessive–compulsive disorder (OCD), bulimia nervosa, panic disorder, and premenstrual dysphoric disorder. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 5 g
CAS Max % 1
UN Number UN3077
Percent Purity 98 to 102%