Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoxymesterone, Acros Organics™

Click to view available options
Quantity:
1g
5g
76-43-7
C20H29FO3
336.447
YLRFCQOZQXIBAB-RBZZARIASA-N
fluoxymesterone,fluoximesterone,fluoxymestrone,halotestin,androfluorene,androfluorone,fluotestin,androsterolo,fluosterone,flusteron
6446
CHEBI:5120
(8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F
76-43-7
C20H29FO3
336.447
YLRFCQOZQXIBAB-RBZZARIASA-N
fluoxymesterone,fluoximesterone,fluoxymestrone,halotestin,androfluorene,androfluorone,fluotestin,androsterolo,fluosterone,flusteron
6446
CHEBI:5120
(8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F
Melting Point 240.0°C
Specific Rotation Condition + 104.00 (20.00°C c=1,EtOH)
Specific Rotation + 104.00
Packaging Glass bottle
Quantity 1g
Solubility Information Solubility in water: negligible. Other solubilities: soluble in puridine, slightly soluble in acetone, chloroform, sparingly soluble in methanol, practically insoluble in ether, benzene, hexanes
Formula Weight 336.45
Chemical Name or Material Fluoxymesterone