Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoxymesterone, Thermo Scientific Chemicals

Catalog Number AC124590050
Click to view available options
Quantity:
5 g
76-43-7
C20H29FO3
336.447
YLRFCQOZQXIBAB-RBZZARIASA-N
fluoxymesterone, fluoximesterone, fluoxymestrone, halotestin, androfluorene, androfluorone, fluotestin, androsterolo, fluosterone, flusteron
6446
CHEBI:5120
(8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F
76-43-7
C20H29FO3
336.447
YLRFCQOZQXIBAB-RBZZARIASA-N
fluoxymesterone, fluoximesterone, fluoxymestrone, halotestin, androfluorene, androfluorone, fluotestin, androsterolo, fluosterone, flusteron
6446
CHEBI:5120
(8S,9R,10S,11S,13S,14S,17S)-9-fluoro-11,17-dihydroxy-10,13,17-trimethyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
CC12CCC(=O)C=C1CCC3C2(C(CC4(C3CCC4(C)O)C)O)F

Fluoxymesterone, CAS # 76-43-7, is a potent, orally administered, synthetic steroid with androgenic activity. It is used in the treatment of male hypogonadism.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Fluoxymesterone is a white, odorless, crystalline powder, soluble in pyridine
  • It is an orally-administered synthetic steroid that is about 5-10 times more potent than male steroid, testosterone
  • It is metabolized in the liver, and less than 5% of this drug is excreted from the body unchanged
  • Adverse effects of fluoxymesterone include sodium and water retention leading to edema, acne, cardiovascular toxicity, and hepatotoxicity

Applications

  • Fluoxymesterone has been found to competitively inhibit prolactin or estrogen receptors in tumorigenic breast cells, and is therefore administered in the treatment of breast cancer
  • In males, this compound is used as a replacement in conditions associated with symptoms of deficiency or absence of endogenous testosterone
TRUSTED_SUSTAINABILITY
Melting Point 240.0°C
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition + 104.00 (20.00°C c=1,EtOH)
Specific Rotation + 104.00
Solubility Information Solubility in water: negligible. Other solubilities: soluble in puridine, slightly soluble in acetone, chloroform, sparingly soluble in methanol, practically insoluble in ether, benzene, hexanes
Formula Weight 336.45
Chemical Name or Material Fluoxymesterone

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit