Learn More
Medchemexpress LLC HY-B1904 10mg Medchemexpress, Fluphenazine decanoate CAS:5002-47-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB190410MG
Encompass_Preferred
Medchemexpress, HY-B1904 10mg Fluphenazine decanoate CAS:5002-47-1 Fluphenazine decanoate is a long-acting phenothiazine neuroleptic that used to treat schizophrenia. Fluphenazine decanoate is also a high and continuous dopamine D2 receptor blocker[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0171