Learn More
Selleck Chemical LLCSupplier Diversity Partner Flupirtine maleate 25mg 75507-68-5  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S133425MG
Flupirtine maleate is the salt form of Flupirtine, which is a centrally acting non-opioid analgesia, is a selective neuronal potassium channel opener that also has NMDA receptor antagonist properties.
Catalog No. 50-652-2