Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Fluprostenol, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1mg
This item is not returnable. View return policy
54276-17-4
C23H29F3O6
458.474
WWSWYXNVCBLWNZ-BVJIBDCISA-N
+-fluprostenol
123789850
7-[(1R,2S,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-4-[3-(trifluoromethyl)phenoxy]but-1-enyl]cyclopentyl]hept-5-enoic acid
C1C(C(C(C1O)C=CC(COC2=CC=CC(=C2)C(F)(F)F)O)CC=CCCCC(=O)O)O
This item is not returnable. View return policy
54276-17-4
C23H29F3O6
458.474
WWSWYXNVCBLWNZ-BVJIBDCISA-N
+-fluprostenol
123789850
7-[(1R,2S,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-4-[3-(trifluoromethyl)phenoxy]but-1-enyl]cyclopentyl]hept-5-enoic acid
C1C(C(C(C1O)C=CC(COC2=CC=CC(=C2)C(F)(F)F)O)CC=CCCCC(=O)O)O

High affinity FP prostaglandin receptor agonist

Prostaglandin F2α (FP) receptor agonist (Ki = 49.9 nM; EC50 = 2.4 nM). Stimulates intracellular calcium mobilization in cloned human ocular FP receptors, rat A7r5 cells and mouse 3T3 cells (EC50 values are 17.5, 19.1 and 37.3 nM respectively). Analog of prostaglandin F2α. Isopropyl ester travaprost (Cat. No. 4218) also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 1mg
Formula Weight 458.47
Percent Purity >95%
Chemical Name or Material (+)-Fluprostenol

Recommended Storage : Store at -20°C