Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flurbiprofen, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
5104-49-4
C15H13FO2
244.27
SYTBZMRGLBWNTM-UHFFFAOYNA-N
2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
5104-49-4
C15H13FO2
244.27
SYTBZMRGLBWNTM-UHFFFAOYNA-N
2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

F1128, 5104-49-4, C15H13FO2

Spectrum™ Flurbiprofen, USP is a non-steroidal anti-inflammatory drug used to treat pain and inflammation. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Melting Range 114°-117°C
Loss on Drying 0.005
Quantity 500 g
CAS Max % 1
UN Number UN2811
Percent Purity 99 to 100.5%