Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flurizan, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
51543-40-9
C15H13FO2
244.265
SYTBZMRGLBWNTM-SNVBAGLBSA-N
tarenflurbil,r-flurbiprofen,flurizan,r-2-flurbiprofen,unii-501w00oowa,r-2-flubiprofen,r---flurbiprofen,--2r-2-2-fluorobiphenyl-4-yl propanoic acid,r---2-fluoro-alpha-methyl-4-biphenylacetic acid
92337
CHEBI:38666
(2R)-2-(3-fluoro-4-phenylphenyl)propanoic acid
CC(C1=CC(=C(C=C1)C2=CC=CC=C2)F)C(=O)O
This item is not returnable. View return policy
51543-40-9
C15H13FO2
244.265
SYTBZMRGLBWNTM-SNVBAGLBSA-N
tarenflurbil,r-flurbiprofen,flurizan,r-2-flurbiprofen,unii-501w00oowa,r-2-flubiprofen,r---flurbiprofen,--2r-2-2-fluorobiphenyl-4-yl propanoic acid,r---2-fluoro-alpha-methyl-4-biphenylacetic acid
92337
CHEBI:38666
(2R)-2-(3-fluoro-4-phenylphenyl)propanoic acid
CC(C1=CC(=C(C=C1)C2=CC=CC=C2)F)C(=O)O

γ-secretase inhibitor; lowers Aβ42 levels in vitro

Nonsteroidal anti-inflammatory drug (NSAID) that inhibits γ-secretase activity. Selectively lowers Aβ42 levels in human neuroglioma cells; displays no effect on Aβ40 levels at a concentration of 100 μM.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 244.26
Percent Purity >99%
Chemical Name or Material Flurizan

Recommended Storage : Store at Room Temperature