Learn More
Flushing Oil
Essential for high vacuum pump maintenance.
$48.09
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity & Availability | |||||
S41456
![]() ![]() |
Hyvac Products Inc![]() 93060002 |
1 gal. |
Each for $48.09
|
|
Description
- Safely remove sediment and contamination from high vacuum pumps
- One or two flushings clean residue and protect pump
- Pump can be refilled immediately after flushing
- 1 gal. size
Use for flushing only. Should not be used for high vacuum operation.