Learn More
FM KRYSTALL FOOD-GRADE PENETRA DFS Item
FM KRYSTALL is a non-viscous penetrating lubricant and cleaner specially developed for use in machinery used in the food and beverage processing and packaging industry. It is based on careful blend of fluids and select additives chosen for their ability to meet the stringent requirements of food and beverage industry. Registered by NSF (Class H1) for use where there is potential for incidental food contact. Produced according to FLT Quality Standards, in facilities where HACCP audit and Good Manufacturing Practice have been implemented and form part of the quality management systems ISO 9001
Catalog No. NC1474790
Please call customer service for current pricing. 1-800-766-7000