Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMN1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549490

Catalog No. PA549490

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
Specifications

Specifications

FMN1
Polyclonal
PBS with 0.09% sodium azide
Q68DA7
Formin-1, Limb deformity protein homolog, FMN1, FMN, LD
Rabbit
IgG
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
342184
Western Blot
Unconjugated
FMN1
Liquid
FMN1
KLH conjugated synthetic peptide between 270-304 amino acids from the Central region of human FMN1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit