Learn More
Aviva Systems Biology Corp FMNL2 Antibody - N-terminal region (ARP47702_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47702_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FMNL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FMNL2. Alternate names: FHOD2, FLJ37546ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-59644