Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FMO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA593278
Description
Immunogen sequence: KPTLAIIGLI KPLGSMIPTG ETQARWAVRV LKGVNKLPPP SVMIEEINAR KENKPSWFGL CYCKALQSDY ITYIDELLTY INAKPNLFSM LLTDPHLALT VFFGPCSPYQ FRLTGPGKWE GARNAIMTQW DRTFKVIKAR VVQESPSPFE SFLKVFSFLA LLVAIFLIFL; Positive Samples: Mouse liver; Cellular Location: Endoplasmic reticulum membrane, Microsome membrane
Metabolic N-oxidation of the diet-derived amino-trimethylamine (TMA) is mediated by flavin-containing monooxygenase and is subject to an inherited FMO3 polymorphism in man resulting in a small subpopulation with reduced TMA N-oxidation capacity resulting in fish odor syndrome Trimethylaminuria. Three forms of the enzyme, FMO1 found in fetal liver, FMO2 found in adult liver, and FMO3 are encoded by genes clustered in the 1q23-q25 region. Flavin-containing monooxygenases are NADPH-dependent flavoenzymes that catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics.Specifications
FMO1 | |
Polyclonal | |
Unconjugated | |
FMO1 | |
Fmo1 | |
Recombit fusion protein containing a sequence corresponding to amino acids 363-532 of human FMO1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Western Blot | |
3.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P50285, Q01740 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14261, 2326 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title