Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMO1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FMO1
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093839
View Documents
Novus Biologicals
NBP304991100UL
100 μg Each for $320.71
Add to cart
 
NB093840
View Documents
Novus Biologicals
NBP30499120UL
20 μg Each for $138.37
Add to cart
 
Description

Description

FMO1 Polyclonal antibody specifically detects FMO1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

FMO1
Unconjugated
Rabbit
Human, Mouse
dimethylaniline monooxygenase [N-oxide-forming] 1, Dimethylaniline oxidase 1, EC 1.14.13.8, Fetal hepatic flavin-containing monooxygenase 1, flavin containing monooxygenase 1, Flavin-containing monooxygenase 1 (fetal liver), FMO 1
Recombinant fusion protein containing a sequence corresponding to amino acids 363-532 of human FMO1 (NP_002012.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
2326
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit