Learn More
Invitrogen™ FMO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595318
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat lung tissue, mouse lung tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The flavin-containing monooxygenases are NADPH-dependent enzymes that catalyze the oxidation of many drugs and xenobiotics. In most mammals, there is a flavin-containing monooxygenase that catalyzes the N-oxidation of some primary alkylamines through an N-hydroxylamine intermediate. However, in humans, this enzyme is truncated and is probably rapidly degraded. The protein encoded by this gene represents the truncated form and apparently has no catalytic activity. A functional allele found in African Americans has been reported, but no sequence evidence has been deposited to support the finding. This gene is found in a cluster with the FMO1, FMO3, and FMO4 genes on chromosome 1.
Specifications
FMO2 | |
Polyclonal | |
Unconjugated | |
Fmo2 | |
2310008D08Rik; 2310042I22Rik; AW107733; Dimethylaniline monooxygenase [N-oxide-forming] 2; dimethylaniline oxidase 2; flavin containing dimethylaniline monoxygenase 2; flavin containing monooxygenase 2; flavin containing monooxygenase 2 (non-functional); FMO 1B1; FMO 2; FMO, pulmonary; FMO1B1; FMO2; Pulmonary flavin-containing monooxygenase 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2327, 246245, 55990 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
Q6IRI9, Q8K2I3, Q99518 | |
Fmo2 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human FMO2 (78-115aa FPNFLHNSKLLEYFRIFAKKFDLLKYIQFQTTVLSVRK). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.