Learn More
FMO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 168611AP
Catalog No. 50-173-1225
Add to cart
Description
Description
The flavin-containing monooxygenases are NADPH-dependent enzymes that catalyze the oxidation of many drugs and xenobiotics. In most mammals, there is a flavin-containing monooxygenase that catalyzes the N-oxidation of some primary alkylamines through an N-hydroxylamine intermediate. However, in humans, this enzyme is truncated and is probably rapidly degraded. The protein encoded by this gene represents the truncated form and apparently has no catalytic activity. A functional allele found in African Americans has been reported, but no sequence evidence has been deposited to support the finding. This gene is found in a cluster with the FMO1, FMO3, and FMO4 genes on chromosome 1.
Specifications
Specifications
FMO2
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6IRI9, Q8K2I3, Q99518
Dimethylaniline oxidase 2, FMO 1B1, FMO 2, FMO1B1, FMO2
Rabbit
IgG
150 μL
-20°C
Primary
2327, 246245, 55990
Western Blot
Unconjugated
FMO2
Liquid
Fmo2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat