Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMO3 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI3H1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270755AF700

Catalog No. N270755A700


Add to cart

Description

Description

FMO3 Monoclonal antibody specifically detects FMO3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

FMO3
OTI3H1
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
FMO3
Recombinant protein fragment corresponding to amino acids 1-291 of human FMO3 (NP_008825) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
dimethylaniline monooxygenase [N-oxide-forming] 3, Dimethylaniline oxidase 3, dJ127D3.1, EC 1.14.13.8, flavin containing monooxygenase 3, FMO 3, FMO form 2, FMO II, FMOII, Hepatic flavin-containing monooxygenase 3, hepatic flavin-containing monooxygenase-3, MGC34400, TMAU
Mouse
IgG1
0.1 mL
Primary
2328.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit