Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FMO3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | FMO3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP182415
|
Novus Biologicals
NBP182415 |
100 μL |
Each of 1 for $436.00
|
|
Description
FMO3 Polyclonal specifically detects FMO3 in Mouse samples. It is validated for Western Blot.Specifications
FMO3 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
dimethylaniline monooxygenase [N-oxide-forming] 3, Dimethylaniline oxidase 3, dJ127D3.1, EC 1.14.13.8, flavin containing monooxygenase 3, FMO 3, FMO form 2, FMO II, FMOII, Hepatic flavin-containing monooxygenase 3, hepatic flavin-containing monooxygenase-3, MGC34400, TMAU | |
FMO3 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_032056 | |
2328 | |
Synthetic peptide towards Fmo3. Peptide sequence KPNVKEFTETSAVFEDGTMFEAIDCVIFATGYGYAYPFLDDSIIKSRNNE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title