Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen FMO4
Concentration 0.2 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-0197
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
153941AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

This antibody, generated by N-330 amino-acids of FMO4, can recognize the 2 variants and also cross-react with other FMO proteins.

FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
Specifications

Specifications

FMO4
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
IgG
-20°C
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit