Learn More
FMO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153941AP
Catalog No. 50-173-0197
Add to cart
Description
Description
This antibody, generated by N-330 amino-acids of FMO4, can recognize the 2 variants and also cross-react with other FMO proteins.
FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
Specifications
Specifications
FMO4
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P31512, Q8K4B7, Q8VHG0
Dimethylaniline oxidase 4, FMO 4, FMO2, FMO4
Rabbit
IgG
150 μL
-20°C
Primary
226564, 2329, 246247
Western Blot
Unconjugated
FMO4
Liquid
Fmo4
FMO4 Fusion Protein Ag7590
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat