Learn More
FMO5 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00002330T01
Description
Western Blotting
Specifications
Western Blot | |
FMO5 293T Cell Transient Overexpression Lysate(Denatured) | |
31.46 | |
pCMV-FMO5 full-length | |
100 μL | |
Human |
293T | |
BC035687.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-FMO5 antibody (H00002330-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |