Learn More
Medchemexpress LLC Fmoc-3VVD-OH (Standard) | 863971-44-2 | MFCD28386982 | 98% | C36H51N3O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY78921R5MG
Encompass_Preferred
Fmoc-3VVD-OH (Standard) is the analytical standard of Fmoc-3VVD-OH. It is intended for research and analytical applications and should be stored at -20°C. Fmoc-3VVD-OH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).
  • Analytical standard
  • Intended for research and analytical applications
  • Functions as a cleavable ADC linker
  • Used in the synthesis of antibody-drug conjugates
  • Categorized under reference standards and ADC linkers
  • Relevant to antibody-drug conjugate/ADC pathways
Catalog No. 50-004-32072