Learn More
Aat BioquestSupplier Diversity Partner FMOC-Asp(5/6-TAMRA)-OH  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 5005
FMOC-Asp(5/6-TAMRA)-OH is a building block for in-sequence peptide Asp labeling by TAMRA. TAMRA is one of the most common donor dyes for preparing FRET peptides (often paired with FAM or TQ3).
Catalog No. 50-195-3247