Learn More
Aat BioquestSupplier Diversity Partner FMOC-Asp(TQ3)-OH  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 5208
FMOC-Asp(TQ3)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 3 (TQ3).
Catalog No. 50-195-3275