Learn More
Medchemexpress LLC Fmoc-Bip(4,4')-OH | 199110-64-0 | 99.8% | 463.52 | 1 G

Supplier: Medchemexpress LLC HYW0108361G
Fmoc-Bip(4,4')-OH is an alanine derivative recognized for its benefits as an ergogenic dietary substance. It is used commercially as an ergogenic supplement, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage.
- Alanine derivative
- Used as ergogenic supplement
- Influences anabolic hormone secretion
- Supplies fuel during exercise
- Improves mental performance during stress-related tasks
- Prevents exercise-induced muscle damage
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.