Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Na-Fmoc-Nw-(2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl)-D-arginine, ≥ 99.5% (Chiral purity), CAS#: 187618-60-6, 25G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0288225G
Na-Fmoc-Nw-(2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl)-D-arginine, ≥ 99.5% (Chiral purity), CAS#: 187618-60-6, 25G. Hazardous Material: NO. MDL #: MFCD00237010. Molecular Formula: C34H40N4O7S. Molecular Weight: 648.77. Appearance: White powder. Optical Rotation: [a]D25 = +4.5 ± 1.5º (C=1 in DMF). Storage Temperature: Store at 0-8 °C.
Catalog No. 50-496-766