Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMOC-D-β-phenylalanine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
209252-15-3
C24H21NO4
387.44
MFCD01311759
PTSLRPMRTOVHAB-QFIPXVFZSA-N
s-n-fmoc-3-amino-3-phenylpropanoic acid, fmoc-beta-s-3-phenylalanine, s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-beta-d-phe-oh, fmoc-s-phenylalanine, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid, fmoc-d-beta-phenylalanine, s-n-fmoc-3-amino-3-phenylpropanoicacid, fmoc-?-phe-oh
7006554
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
209252-15-3
C24H21NO4
387.44
MFCD01311759
PTSLRPMRTOVHAB-QFIPXVFZSA-N
s-n-fmoc-3-amino-3-phenylpropanoic acid, fmoc-beta-s-3-phenylalanine, s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-beta-d-phe-oh, fmoc-s-phenylalanine, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid, fmoc-d-beta-phenylalanine, s-n-fmoc-3-amino-3-phenylpropanoicacid, fmoc-?-phe-oh
7006554
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Specific Rotation -21° to -25° (20°C, 589nm) (c=1.1, DMF)
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 186.0°C to 194.0°C
Quantity 1g
Color White
Assay Percent Range 94% min. (HPLC)
Formula Weight 387.44
Physical Form Powder
Percent Purity 95%
Chemical Name or Material FMOC-D-β-phenylalanine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning