Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Na-Fmoc-Nin-Boc-D-tryptophan, ≥ 98% (HPLC), CAS#: 163619-04-3, 25G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0248425G
Na-Fmoc-Nin-Boc-D-tryptophan, ≥ 98% (HPLC), CAS#: 163619-04-3, 25G. Hazardous Material: NO. MDL #: MFCD00153367. Molecular Formula: C31H30N2O6. Molecular Weight: 526.6. Appearance: White to off-white powder. Optical Rotation: [a]D20 = +20 ± 4º (C=1 in DMF). Storage Temperature: Store at 0-8 °C.
Catalog No. 50-495-786