Learn More
Aat BioquestSupplier Diversity Partner FMOC-Dap(Boc)-OH *CAS 162558-25-0* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 5110
FMOC-Dap(Boc)-OH is a common peptide building block. As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.
Catalog No. 50-195-3272