Learn More
Aat BioquestSupplier Diversity Partner FMOC-Glu(EDANS)-OH *CAS 193475-66-0*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 5009
FMOC-Glu(EDANS)-OH is a building block for in-sequence peptide Glu labeling by EDANS. EDANS is one of the most common donor dyes for preparing FRET peptides (often paired with Dabcyl).
Catalog No. 50-195-3250