Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Fmoc-L-glutamic acid α-allyl ester,144120-54-7,MFCD00467718,5G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 036395G
Molecular Formula C23H23NO6; Purity Limit ≥ 98% (HPLC); Molecular Weight 409.43; Melting Point 118-133 °C; Store at 0-8 °C; Synonyms: Fmoc-L-Glu-OAll; Allyl esters are useful carboxy protecting groups in the synthesis of various cyclic peptides glycopeptides. Allyl esters are cleaved quantitatively under mild conditions using Pd(0) catalyst.
Catalog No. 50-498-094