Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Fmoc-L-glutamic acid γ-tert-butyl ester hydrate,204251-24-1, 71989-18-9,MFCD00150485,25G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0241325G
Molecular Formula C24H27NO6·H2O, Purity Limit ≥ 99.7% (Chiral HPLC), Molecular Weight 443.49, Melting Point 80 - 100 °C, Storage Temp: Store at 0-8 °C, Synonyms: Fmoc-L-Glu(OtBu)-OH hydrate, Fmoc-L-glutamic acid 5-tert-butyl ester hydrate
Catalog No. 50-495-550