Learn More
Aat BioquestSupplier Diversity Partner FMOC-Glu(TQ2)-OH  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 5236
FMOC-Glu(TQ2)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 2 (TQ2).
Catalog No. 50-195-3277