Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Fmoc-glycine,29022-11-5,MFCD00037140,100G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 02416100G
Molecular Formula C17H15NO4, Purity Limit ≥ 99% (HPLC), Molecular Weight 297.30, Melting Point 165 - 185 °C, Storage Temp: Store at 0 - 8 °C, Synonyms: Fmoc-Gly-OH
Catalog No. 50-495-562