Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Fmoc-L-trans-4-hydroxyproline, 88050-17-3, MFCD00151929, 25g DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0242325G
Molecular Formula C20H19NO5, Purity Limit ≥ 99% (HPLC), Molecular Weight 353.4, Melting Point 186-196 °C, Synonyms: Fmoc-L-Hyp-OH
Catalog No. 50-495-583