Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMOC-L-Norleucine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
77284-32-3
C21H23NO4
353.42
VCFCFPNRQDANPN-IBGZPJMESA-N
fmoc-nle-oh,fmoc-l-norleucine,n-fmoc-l-norleucine,s-2-fmoc-amino caproic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,fmoc-norleucine,fmocnleoh,fmoc-nle
7009636
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
77284-32-3
C21H23NO4
353.42
VCFCFPNRQDANPN-IBGZPJMESA-N
fmoc-nle-oh,fmoc-l-norleucine,n-fmoc-l-norleucine,s-2-fmoc-amino caproic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,fmoc-norleucine,fmocnleoh,fmoc-nle
7009636
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Specific Rotation -19
Melting Point 138°C to 142°C
Color White
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 353.42
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material FMOC-L-Norleucine

GHS Signal Word: Warning