Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Na-Fmoc-Ne-methyltrityl-L-lysine, ≥ 99.8% (HPLC, Chiral purity), CAS#: 167393-62-6, 5G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 037205G
Na-Fmoc-Ne-methyltrityl-L-lysine, ≥ 99.8% (HPLC, Chiral purity), CAS#: 167393-62-6, 5G. Hazardous Material: NO. MDL #: MFCD00237166. Molecular Formula: C41H40N2O4. Molecular Weight: 624.79. Melting Point: 135-148 °C. Appearance: White to off-white powder. Optical Rotation: [a]D20 = +4 ± 1º (C=1 in THF). Storage Temperature: Store at 0-8 °C.
Catalog No. 50-498-214