Learn More
Medchemexpress LLC Fmoc-Lys-OMe HCl | 847658-45-1 | MFCD08275774 | 99.8% | 418.91 | C22H27ClN2O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0190285G
Encompass_Preferred
Fmoc-Lys-OMe HCl is an Fmoc-protected lysine methyl ester hydrochloride used as a building block in peptide synthesis and other amino-acid derivative chemistry. Supplied as the hydrochloride salt with high reported purity, it is intended for research use in laboratory-scale synthesis and modification workflows.
  • Fmoc-protected amino acid suitable for peptide coupling.
  • Methyl ester provides orthogonal protection for side-chain modification.
  • Hydrochloride salt enhances stability and handling.
  • High purity for reliable synthetic performance.
  • Soluble in common organic solvents for standard protocols.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-003-00718